Learn More
Enzo Life Sciences Trichostatin A (5mg). CAS: 58880-19-6
SDP
Supplier:  Enzo Life Sciences BMLGR3090005
Encompass_Preferred
Trichostatin A (TSA) is a potent and reversible inhibitor of histone deacetylases. In HeLa cells, TSA blocked cell cycle progression at G1 and induced a 12-fold increase in intracellular levels of gelsolin. In cells latently infected with HIV-1, TSA induced the transcriptional activation of the HIV-1 promoter, which resulted in a marked increase in virus production. In NIH 3T3 cells, TSA induced reversion of oncogenic ras-transformed cells to a normal morphology. In Jurkat cells, TSA inhibited IL-2 gene expression (IC50=73nM) and displayed immunosuppressive activity in a mouse model. Induces increased acetylation of GATA4, a cardiac-specific transcription factor and increases cardiac muscle cell differentiation. Trichostatin A is a useful tool for induction of hyperacetylation of cellular histones and for further elucidation of their role in gene expression. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO, dimethyl formamide, acetonitrile, methanol or 100% ethanol.
Catalog No. 50-201-0960