Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Trichostatin A 98.0+%, TCI America™
SDP

Catalog No. T363310MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T363310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T363310MG Supplier TCI America Supplier No. T363310MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 122292-85-7
Molecular Formula C17H22N2O3
Molecular Weight (g/mol) 302.374
InChI Key RTKIYFITIVXBLE-LEJRBOCMSA-N
Synonym [S-(E,E)]-(-)-7-[4-(Dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide
PubChem CID 13507024
IUPAC Name (2E,4E,6S)-7-[4-(dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxohepta-2,4-dienamide
SMILES CC(C=C(C)C=CC(=O)NO)C(=O)C1=CC=C(C=C1)N(C)C

Specifications

Color White-Yellow
Quantity 10 mg
Formula Weight 302.37
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Trichostatin A
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.