Learn More
Selleck Chemical LLC Trichostatin A TSA-2mg
SDP
Catalog No. 502389588
Encompass_Preferred
Trichostatin A (TSA) is an HDAC inhibitor with IC50 of ~1.8 nM in cell-free assays.
Catalog No. 50-238-9588 Supplier Selleck Chemical LLC Supplier No. S10452MG
Trichostatin A (TSA) is an HDAC inhibitor with IC50 of ~1.8 nM in cell-free assays.