Learn More
Newcomer Supply Trichrome, McLetchie, Aniline Blue Stain Kit, 250 ml kit
SDP
Catalog No. NC2102300
Encompass
Newcomer Supply Trichrome, McLetchie, Aniline Blue Stain Kit  procedure is used for the differential demonstration of collagen and muscle fibers.  This modified trichrome protocol provides time efficient results without the use of Bouin Fluid or a hematoxylin nuclear stain.
This item is not returnable. View return policy
Catalog No. NC2102300 Supplier Newcomer Supply Supplier No. 9177A
Newcomer Supply Trichrome, McLetchie, Aniline Blue Stain Kit  procedure is used for the differential demonstration of collagen and muscle fibers.  This modified trichrome protocol provides time efficient results without the use of Bouin Fluid or a hematoxylin nuclear stain.