Learn More
Newcomer Supply Trichrome, Gomori One-Step, Aniline Blue Stain Kit, 250 ml kit
SDP
Catalog No. NC2102298
Encompass
Newcomer Supply Trichrome, Gomori One-Step, Aniline Blue Stain Kit procedure, with included microwave modification, uses a one-step solution combining a plasma stain and a connective tissue stain to differentially demonstrate collagen and muscle fibers.
This item is not returnable. View return policy
Catalog No. NC2102298 Supplier Newcomer Supply Supplier No. 9176B
Newcomer Supply Trichrome, Gomori One-Step, Aniline Blue Stain Kit procedure, with included microwave modification, uses a one-step solution combining a plasma stain and a connective tissue stain to differentially demonstrate collagen and muscle fibers.