Learn More
Newcomer Supply Trichrome, Gomori One-Step, Fast Green Stain Kit, 250 ml kit
SDP
Catalog No. NC2102296
Encompass
Newcomer Supply Trichrome, Gomori One-Step, Fast Green Stain Kit procedure, with included microwave modification, uses a one-step solution combining a plasma stain and a connective tissue stain to differentially demonstrate collagen and muscle fibers.
This item is not returnable. View return policy
Catalog No. NC2102296 Supplier Newcomer Supply Supplier No. 9175A
Newcomer Supply Trichrome, Gomori One-Step, Fast Green Stain Kit procedure, with included microwave modification, uses a one-step solution combining a plasma stain and a connective tissue stain to differentially demonstrate collagen and muscle fibers.