Learn More
Newcomer Supply Trichrome, Masson, Fast Green Stain Kit, 250ml kit
SDP
Catalog No. NC2102302
Encompass
Newcomer Supply Trichrome, Masson, Fast Green Stain Kit procedure, with included microwave modification, is used to differentially demonstrate connective tissue elements, collagen and muscle fibers.
This item is not returnable. View return policy
Catalog No. NC2102302 Supplier Newcomer Supply Supplier No. 9180A
Newcomer Supply Trichrome, Masson, Fast Green Stain Kit procedure, with included microwave modification, is used to differentially demonstrate connective tissue elements, collagen and muscle fibers.