Learn More
Volu Sol Trichrome Blue Stain (Ryan Modification), 16 oz / 500 mL, Volu-Sol™
SDP
Catalog No. 50420837 Shop All Volu Sol Products
Encompass_Preferred
Trichrome Blue, recommended for the detection of microsporidia in urine, stool, and nasopharyngeal specimens
Catalog No. 50-420-837 Supplier Volu Sol Supplier No. VBT016
Trichrome Blue, recommended for the detection of microsporidia in urine, stool, and nasopharyngeal specimens