Learn More
Newcomer Supply Trichrome, Multi-Tissue Control Slides, 10/set
SDP
Catalog No. NC1955685
Encompass
Newcomer Supply Trichrome, Multi-Tissue Control Slides, use a combination of tissue sources for the positive histochemical staining of connective tissue and to differentially demonstrate collagen and muscle fibers. Trichrome, Multi-Tissue Control Slides contain sections of positive staining liver, positive staining kidney and positive staining uterus.
Catalog No. NC1955685 Supplier Newcomer Supply Supplier No. 4693A
Newcomer Supply Trichrome, Multi-Tissue Control Slides, use a combination of tissue sources for the positive histochemical staining of connective tissue and to differentially demonstrate collagen and muscle fibers. Trichrome, Multi-Tissue Control Slides contain sections of positive staining liver, positive staining kidney and positive staining uterus.