Learn More
Abcam Trichrome Stain Kit, Masson's Trichrome, 100 tests
Catalog No. NC0897059
Encompass_Preferred
The Abcam AB150686-100TEST is a Trichrome Stain Kit designed for histological visualization of collagenous connective tissue fibers. This kit enables differential staining of collagen and other tissue components, making it essential for researchers studying fibrosis and collagen levels. It is optimized for formalin-fixed, paraffin-embedded tissue sections but can also be adapted for frozen sections with optimal conditions. Below are the key features and benefits of this product:
  • Valuable tool for studying fibrosis and collagen levels
  • Stains collagen blue, muscle fibers red, and nuclei blue
  • Includes additional staining for fibrin and erythrocytes
  • Control tissues include lung, uterus, small intestine, and stomach
  • Contains specific stains such as Aniline Blue and Weigert's Hematoxylin
  • Supplies enough reagents for 100 tests
  • Designed for research use only
  • Compatible with human, mouse, and rat tissues
Catalog No. NC0897059 Supplier Abcam Supplier No. AB150686100TEST
The Abcam AB150686-100TEST is a Trichrome Stain Kit designed for histological visualization of collagenous connective tissue fibers. This kit enables differential staining of collagen and other tissue components, making it essential for researchers studying fibrosis and collagen levels. It is optimized for formalin-fixed, paraffin-embedded tissue sections but can also be adapted for frozen sections with optimal conditions. Below are the key features and benefits of this product:
  • Valuable tool for studying fibrosis and collagen levels
  • Stains collagen blue, muscle fibers red, and nuclei blue
  • Includes additional staining for fibrin and erythrocytes
  • Control tissues include lung, uterus, small intestine, and stomach
  • Contains specific stains such as Aniline Blue and Weigert's Hematoxylin
  • Supplies enough reagents for 100 tests
  • Designed for research use only
  • Compatible with human, mouse, and rat tissues