Learn More
Newcomer Supply Trichrome, Gomori One-Step, Aniline Blue Stain Kit, 500 ml kit
SDP
Catalog No. NC9485545
Encompass
Newcomer Supply Trichrome, Gomori One-Step, Aniline Blue Stain Kit procedure, with included microwave modification, uses a one-step solution combining a plasma stain and a connective tissue stain to differentially demonstrate collagen and muscle fibers.
Catalog No. NC9485545 Supplier Newcomer Supply Supplier No. 9176A
Newcomer Supply Trichrome, Gomori One-Step, Aniline Blue Stain Kit procedure, with included microwave modification, uses a one-step solution combining a plasma stain and a connective tissue stain to differentially demonstrate collagen and muscle fibers.