Learn More
Sigma Aldrich Fine Chemicals Biosciences Trichrome Stain (Masson) Kit
Catalog No. NC1792381
Encompass
May be useful in study of diseases of connective tissue and muscle characterized by fibrotic and dystrophic changes. Collagen and muscle fiber stain differentially when treated sequentially with Biebrich Scarlet-Acid Fuchsin, PTA/PMA and Aniline Blue. Cytoplasm and muscle fibers stain red whereas collagen displays blue coloration.
Catalog No. NC1792381 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HT151KT
May be useful in study of diseases of connective tissue and muscle characterized by fibrotic and dystrophic changes. Collagen and muscle fiber stain differentially when treated sequentially with Biebrich Scarlet-Acid Fuchsin, PTA/PMA and Aniline Blue. Cytoplasm and muscle fibers stain red whereas collagen displays blue coloration.