Learn More
Medchemexpress LLC HY-N1127 5mg Medchemexpress, Tricin CAS:520-32-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN11275MG
Encompass_Preferred
Medchemexpress, HY-N1127 5mg Tricin CAS:520-32-1 Tricin is a natural flavonoid present in large amounts in rice bran. Tricin can inhibit human cytomegalovirus (HCMV) replication by inhibiting CDK9. Tricin inhibits the proliferation and invasion of C6 glioma cells via the upregulation of focal-adhesion-finase (FAK)-targeting microRNA-7[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0322