Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tricin 7-O-glucuronide | 32769-02-1 | >98.0% | 506.41 g/mol | C23H22O13 | 5 MG
SDP

Catalog No. 5000263838
Encompass_Preferred

Tricin 7-O-glucuronide is a plant-derived flavonoid glycoside reported from alfalfa and other species. It is used as a research reagent in natural-products, enzymology, and analytical studies where a defined flavonoid glucuronide is required.

  • CAS number 32769-02-1.
  • Chemical formula C23H22O13.
  • Molecular weight 506.41 g/mol.
  • Purity typically >98% (HPLC) according to supplier specifications.
  • Typical pack sizes include 1 mg, 5 mg, and 10 mg vials for research use.
  • Storage: 4°C, protect from light; in solvent store at -80°C for long-term stability.

Catalog No. 50-002-63838 Supplier Medchemexpress LLC Supplier No. HY1118125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tricin 7-O-glucuronide is a plant-derived flavonoid glycoside reported from alfalfa and other species. It is used as a research reagent in natural-products, enzymology, and analytical studies where a defined flavonoid glucuronide is required.

  • CAS number 32769-02-1.
  • Chemical formula C23H22O13.
  • Molecular weight 506.41 g/mol.
  • Purity typically >98% (HPLC) according to supplier specifications.
  • Typical pack sizes include 1 mg, 5 mg, and 10 mg vials for research use.
  • Storage: 4°C, protect from light; in solvent store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.