Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tricin 7-O-glucuronide | 32769-02-1 | 99.1% | 506.41 | 10 MG
SDP

Catalog No. 5000440018
Encompass_Preferred

Tricin 7-O-glucuronide is an Alfalfa (Medicago sativa L.) flavonoid that can be obtained from the leaves of M. tenuifollium G. Forster. It is for research use only.

  • Solid, light yellow to yellow appearance
  • Molecular formula: C23H22O13
  • Classified as a flavonoid, flavone, phenol, and polyphenol
  • Initial source is plants, specifically Medicago sativa L.
  • Solubility in DMSO: ≥ 100 mg/mL
  • Store at 4°C, protected from light, under nitrogen

Catalog No. 50-004-40018 Supplier Medchemexpress LLC Supplier No. HY11181210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tricin 7-O-glucuronide is an Alfalfa (Medicago sativa L.) flavonoid that can be obtained from the leaves of M. tenuifollium G. Forster. It is for research use only.

  • Solid, light yellow to yellow appearance
  • Molecular formula: C23H22O13
  • Classified as a flavonoid, flavone, phenol, and polyphenol
  • Initial source is plants, specifically Medicago sativa L.
  • Solubility in DMSO: ≥ 100 mg/mL
  • Store at 4°C, protected from light, under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.