Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Tricine

Catalog No. 04807410 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-807-410 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-807-410 Supplier MP Biomedicals™ Supplier No. 04807410
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tricine is for use in electrophoresis buffers.

  • Tricine is used as buffer component for separation of low molecular weight peptides.
  • Tricine can be used in cryopreservation medium for the preservation of tissues and organs.
  • Cryopreservation depends on the physical and chemical characteristics of the preservation medium used.
  • The pH values and pK values for tricine/DMSO mixtures has been reported down to -20°C.
  • Tricine has been found to be an efficient scavenger of hydroxyl radicals in a study of radiation-induced membrane damage.
  • Tricine is typically the buffer of choice in SDS-PAGE systems when separating proteins in the range of 1 to 100 kDa.
A buffer may be prepared by titrating with sodium hydroxide to the desired pH, using about a half-equivalent of NaOH.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tricine
CAS 5704-04-1
Color Clear, colorless
Description N-Tris-[hydroxymethyl]methylglycine, N-[2-Hydroxy-1,1-bis(hydroxymethyl) ethyl] glycine
For Use With (Application) Electrophoresis
pH 4.5 to 5.5
Physical Form Powder
Quantity 25 g
Solution Type Buffer
Solubility Information Very soluble in water (25% w/v - clear, colorless solution).
Molecular Weight (g/mol) 179.17
Percent Purity ≥99%
Grade Electrophoresis
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.