Learn More
Medchemexpress LLC Tricine | 5704-04-1 | 99.9% | 1 ML
SDP
Catalog No. 5000409988
Encompass_Preferred
Tricine is a biochemical reagent widely utilized as a buffer in various life science applications. It plays a crucial role in protein electrophoresis, nucleic acid analysis, and immunology by effectively adjusting pH and stabilizing chemical reactions. Furthermore, Tricine finds application in medical research, including antibody detection and drug screening, making it suitable for use as a biological material or organic compound in diverse scientific investigations.
  • Widely used as a buffer in protein electrophoresis, nucleic acid analysis, and immunology.
  • Helps adjust pH.
  • Stabilizes chemical reactions.
  • Can be used in research for antibody detection and drug screening.
  • Biochemical reagent for life science-related research.
Catalog No. 50-004-09988 Supplier Medchemexpress LLC Supplier No. HYD086010MM/1ML
Tricine is a biochemical reagent widely utilized as a buffer in various life science applications. It plays a crucial role in protein electrophoresis, nucleic acid analysis, and immunology by effectively adjusting pH and stabilizing chemical reactions. Furthermore, Tricine finds application in medical research, including antibody detection and drug screening, making it suitable for use as a biological material or organic compound in diverse scientific investigations.
  • Widely used as a buffer in protein electrophoresis, nucleic acid analysis, and immunology.
  • Helps adjust pH.
  • Stabilizes chemical reactions.
  • Can be used in research for antibody detection and drug screening.
  • Biochemical reagent for life science-related research.