Learn More
Medchemexpress LLC Tricine | 5704-04-1 | 99.9% | 10 G
SDP
Catalog No. 5000410409
Encompass_Preferred
Tricine is widely used as a buffer in protein electrophoresis, nucleic acid analysis, and immunology, helping to adjust pH and stabilize chemical reactions. It can also be used in research in certain medical fields, such as antibody detection and drug screening. Tricine is a biochemical reagent that can be used as a biological material or organic compound for life science-related research.
  • Used as a buffer in protein electrophoresis, nucleic acid analysis, and immunology
  • Helps adjust pH and stabilize chemical reactions
  • Applicable in antibody detection and drug screening
  • Biochemical reagent for life science-related research
Catalog No. 50-004-10409 Supplier Medchemexpress LLC Supplier No. HYD086010G
Tricine is widely used as a buffer in protein electrophoresis, nucleic acid analysis, and immunology, helping to adjust pH and stabilize chemical reactions. It can also be used in research in certain medical fields, such as antibody detection and drug screening. Tricine is a biochemical reagent that can be used as a biological material or organic compound for life science-related research.
  • Used as a buffer in protein electrophoresis, nucleic acid analysis, and immunology
  • Helps adjust pH and stabilize chemical reactions
  • Applicable in antibody detection and drug screening
  • Biochemical reagent for life science-related research