Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Tricine, 98%, for biochemistry

Catalog No. AC172641000
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC172641000 100 g Plastic bottle
AC172640250 25 g Plastic bottle
AC172642500 250 g Plastic bottle
AC172640010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC172641000 Supplier Thermo Scientific Chemicals Supplier No. 172641000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4-1-5704
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
Synonym N-[Tris(hydroxymethyl)methyl]glycine, N-(2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl)glycine
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Absorbance 0.05 max. at 250nm
Absorbance Conditions (5% aq. soln.) (1 cm cell)
Melting Point 180.0°C to 184.0°C
Color White
pH 4.8 to 5.4 (0.5M aq. soln. at 20°C)
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 98%
Infrared Spectrum Authentic
Loss on Drying 1% max. (120°C)
Linear Formula (HOCH2)3CNHCH2CO2H
Packaging Plastic bottle
Merck Index 15, 9816
Solubility Information Solubility in water: soluble.
Formula Weight 179.17
Percent Purity 98%
Grade Biochemical
Chemical Name or Material Tricine, 98%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 227-193-6
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.