Learn More
Bio Rad Laboratories TRICINE SAMPLE BUFFER
Catalog No. 509012450
Encompass
The zinc stain and destain kit is a reversible negative stain for proteins in polyacrylamide gels containing SDS. The kit includes 125 ml 10x imidazole (stain solution A), 125 ml 10x zinc sulfate (stain solution B), and 125 ml 10x zinc dest
Catalog No. 50-901-2450 Supplier Bio Rad Laboratories Supplier No. 1610739
The zinc stain and destain kit is a reversible negative stain for proteins in polyacrylamide gels containing SDS. The kit includes 125 ml 10x imidazole (stain solution A), 125 ml 10x zinc sulfate (stain solution B), and 125 ml 10x zinc dest