Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Tricine-SDS Sample Buffer 2X Solution, 50 ML
SDP

Catalog No. 501978242
Encompass_Preferred

2X Formulation:

TRIS HCI: 900 mM
SDS: 8%
Glycerol: 20%
Coomassie Blue: 0.01%
Phenol Red: 0.01%

Catalog No. 50-197-8242 Supplier Research Products International Corp Supplier No. T2720050.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2X Formulation:

TRIS HCI: 900 mM
SDS: 8%
Glycerol: 20%
Coomassie Blue: 0.01%
Phenol Red: 0.01%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.