Learn More
Alkali Scientific Tricine SDS Sample Buffer 2X Solution, 50 Milliliters
SDP
Catalog No. 5000660350
Encompass_Preferred
Tricine SDS Sample Buffer 2X Solution is formulated to ensure efficient protein separation in SDS-PAGE analysis, especially for low molecular weight proteins. This buffer contains Tricine, which improves the resolution of small proteins, making it an excellent choice for detailed protein profiling. The 50mL bottle provides enough volume for several protein analysis applications, especially in academic and research labs. This buffer aids in sample preparation by maintaining protein denaturation and preventing degradation, which is crucial for obtaining clear, reproducible results in SDS-PAGE gel electrophoresis. It’s widely used in proteomics and molecular biology.
Catalog No. 50-006-60350 Supplier Alkali Scientific Supplier No. CIN002CP
Tricine SDS Sample Buffer 2X Solution is formulated to ensure efficient protein separation in SDS-PAGE analysis, especially for low molecular weight proteins. This buffer contains Tricine, which improves the resolution of small proteins, making it an excellent choice for detailed protein profiling. The 50mL bottle provides enough volume for several protein analysis applications, especially in academic and research labs. This buffer aids in sample preparation by maintaining protein denaturation and preventing degradation, which is crucial for obtaining clear, reproducible results in SDS-PAGE gel electrophoresis. It’s widely used in proteomics and molecular biology.