Learn More
Alkali Scientific Tricine, Ultra Pure [N [Tris (Hydroxymethyl) Methyl] Glycine], 5 Kilograms
SDP
Catalog No. 5000661744
Encompass_Preferred
Tricine, Ultra Pure, is an amino acid derivative used as a high-purity buffering agent in laboratory applications. Available in a 5-kilogram package, this ultra-pure grade tricine is ideal for sensitive biochemical and molecular biology experiments that require precise pH control. Tricine is particularly useful for protein electrophoresis and is often employed in the preparation of buffers for SDS-PAGE and other separation techniques. Its low charge at physiological pH makes it suitable for applications where minimal interference with biomolecules is required. This product is used in advanced protein studies, enzyme assays, and other applications where high purity and stable buffering conditions are essential.
Catalog No. 50-006-61744 Supplier Alkali Scientific Supplier No. RIC004CP
Tricine, Ultra Pure, is an amino acid derivative used as a high-purity buffering agent in laboratory applications. Available in a 5-kilogram package, this ultra-pure grade tricine is ideal for sensitive biochemical and molecular biology experiments that require precise pH control. Tricine is particularly useful for protein electrophoresis and is often employed in the preparation of buffers for SDS-PAGE and other separation techniques. Its low charge at physiological pH makes it suitable for applications where minimal interference with biomolecules is required. This product is used in advanced protein studies, enzyme assays, and other applications where high purity and stable buffering conditions are essential.