Learn More
Medchemexpress LLC Triciribine | 35943-35-2 | C13H16N6O4 | 100 MG
SDP
Catalog No. 5000376546
Encompass_Preferred
Triciribine is a nucleoside analog and purine analog that functions as a DNA synthesis inhibitor. It also inhibits Akt and HIV-1/2 with specific IC50 values. This compound selectively inhibits the phosphorylation and activation of all three Akt isoforms, effectively inhibiting Akt catalytic activity in PC-3 cells at 10 μM.
  • Inhibits DNA synthesis
  • Inhibits Akt phosphorylation and activation
  • Inhibits HIV-1/2
  • Effective at inhibiting catalytic activity of Akt in PC-3 cells
Catalog No. 50-003-76546 Supplier Medchemexpress LLC Supplier No. HY15457100MG
Triciribine is a nucleoside analog and purine analog that functions as a DNA synthesis inhibitor. It also inhibits Akt and HIV-1/2 with specific IC50 values. This compound selectively inhibits the phosphorylation and activation of all three Akt isoforms, effectively inhibiting Akt catalytic activity in PC-3 cells at 10 μM.
  • Inhibits DNA synthesis
  • Inhibits Akt phosphorylation and activation
  • Inhibits HIV-1/2
  • Effective at inhibiting catalytic activity of Akt in PC-3 cells