Learn More
Selleck Chemical LLC Triciribine 50mg 35943-35-2 NSC 154020, VD-0002, vqd-002, API-2
SDP
Catalog No. 501360936
Encompass_Preferred
Triciribine (NSC 154020, VD-0002, vqd-002, API-2) is a DNA synthesis inhibitor, also inhibits Akt in PC3 cell line and HIV-1 in CEM-SS, H9, H9IIIB, U1 cells with IC50 of 130 nM and 20 nM, respectively; does not inhibit PI3K/PDK1; 5000-fold less active in cells lacking adenosine kinase. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0936 Supplier Selleck Chemical LLC Supplier No. S111750MG
Triciribine (NSC 154020, VD-0002, vqd-002, API-2) is a DNA synthesis inhibitor, also inhibits Akt in PC3 cell line and HIV-1 in CEM-SS, H9, H9IIIB, U1 cells with IC50 of 130 nM and 20 nM, respectively; does not inhibit PI3K/PDK1; 5000-fold less active in cells lacking adenosine kinase. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.