Learn More
Enzo Life Sciences Triciribine (5mg). CAS: 35943-35-2
SDP
Catalog No. 502010719
Encompass_Preferred
Triciribine is a synthetic tricyclic nucleoside which acts as a specific inhibitor of the Akt signaling pathway. It selectively inhibits the phosphorylation and activation of Akt1, -2 and -3 but does not inhibit Akt kinase activity nor known upstream Akt activators such as PI 3-Kinase and PDK1. It inhibits cell growth and induces apoptosis preferentially in cells that express aberrant Akt1. In whole cells triciribine is phosphorylated by adenosine kinase which may be necessary for its activity. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-0719 Supplier Enzo Life Sciences Supplier No. BMLEI3320005
Add to Cart
Add to Cart
Triciribine is a synthetic tricyclic nucleoside which acts as a specific inhibitor of the Akt signaling pathway. It selectively inhibits the phosphorylation and activation of Akt1, -2 and -3 but does not inhibit Akt kinase activity nor known upstream Akt activators such as PI 3-Kinase and PDK1. It inhibits cell growth and induces apoptosis preferentially in cells that express aberrant Akt1. In whole cells triciribine is phosphorylated by adenosine kinase which may be necessary for its activity. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.