Learn More
Medchemexpress LLC HY-15457 5mg Medchemexpress, Triciribine CAS:35943-35-2 Purity:>98%
Supplier: Medchemexpress LLC HY154575MG
Medchemexpress, HY-15457 5mg Triciribine CAS:35943-35-2 Triciribine is a DNA synthesis inhibitor, also inhibits Akt and HIV-1/2 with IC50 of 130 nM, and 0.02-0.46 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.