Learn More
Abcam Triciribine, inhibitor of Akt activation, 1MG
Catalog No. 502707113
Encompass_Preferred
MW 320.31 g/mol, Purity >98%. Cell-permeable tricyclic nucleoside that inhibits the phosphorylation, activation and signalling of Akt. Selective over PKC, PKA, SGK and p38 pathways. Induces cell growth arrest and apoptosis in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7113 Supplier Abcam Supplier No. AB1209361MG
MW 320.31 g/mol, Purity >98%. Cell-permeable tricyclic nucleoside that inhibits the phosphorylation, activation and signalling of Akt. Selective over PKC, PKA, SGK and p38 pathways. Induces cell growth arrest and apoptosis in vitro.
The product is subject to the following: Abcam Restricted Use Statement