Learn More
Abcam Triciribine, inhibitor of Akt activation, 5MG
Catalog No. 502707112
Encompass_Preferred
MW 320.31 g/mol, Purity >98%. Cell-permeable tricyclic nucleoside that inhibits the phosphorylation, activation and signalling of Akt. Selective over PKC, PKA, SGK and p38 pathways. Induces cell growth arrest and apoptosis in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7112 Supplier Abcam Supplier No. AB1209365MG
MW 320.31 g/mol, Purity >98%. Cell-permeable tricyclic nucleoside that inhibits the phosphorylation, activation and signalling of Akt. Selective over PKC, PKA, SGK and p38 pathways. Induces cell growth arrest and apoptosis in vitro.
The product is subject to the following: Abcam Restricted Use Statement