Learn More
Medchemexpress LLC Triciribine phosphate | 00-00-0 | ≥99.0% | 400.28 g/mol | C13H17N6O7P | 5 MG
SDP
Catalog No. 5000249006
Encompass_Preferred
Triciribine phosphate is the phosphorylated form of triciribine and is supplied as a purified small-molecule reagent for biochemical and cellular research. It is used to modulate signaling pathways, including inhibition of AKT phosphorylation, and may affect nucleotide synthesis pathways. Refer to the manufacturer's product page and safety data sheet for handling, storage, and safety information.
  • Inhibits AKT phosphorylation and related signaling pathways.
  • High purity suitable for biochemical and cellular assays.
  • Provided as a solid for dissolution in common laboratory solvents.
  • Available in milligram-scale pack sizes for laboratory use.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-49006 Supplier Medchemexpress LLC Supplier No. HY10250A5MG
Triciribine phosphate is the phosphorylated form of triciribine and is supplied as a purified small-molecule reagent for biochemical and cellular research. It is used to modulate signaling pathways, including inhibition of AKT phosphorylation, and may affect nucleotide synthesis pathways. Refer to the manufacturer's product page and safety data sheet for handling, storage, and safety information.
  • Inhibits AKT phosphorylation and related signaling pathways.
  • High purity suitable for biochemical and cellular assays.
  • Provided as a solid for dissolution in common laboratory solvents.
  • Available in milligram-scale pack sizes for laboratory use.
  • Intended for research use only; not for human therapeutic use.