Learn More
TARGETMOL CHEMICALS INC Triciribine phosphate 1MG
SDP
Catalog No. 502878274
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. Triciribine phosphate (VD 002) is a highly selective AKT inhibitor that induces cell cycle arrest and cysteinyl asparagin-dependent apoptosis, inhibits neovascularization, and can be used in the study of leukemia. Purity 99.43%
Catalog No. 50-287-8274 Supplier TARGETMOL CHEMICALS INC Supplier No. T196991MG
Also available in 5 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. Triciribine phosphate (VD 002) is a highly selective AKT inhibitor that induces cell cycle arrest and cysteinyl asparagin-dependent apoptosis, inhibits neovascularization, and can be used in the study of leukemia. Purity 99.43%