Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Triciribine phosphate-d3 | 99.7% | 403.30 | C13H14D3N6O7P | 1 MG
SDP

Catalog No. 5000246341
Encompass_Preferred

Triciribine phosphate-d3 is the deuterated isotopologue of triciribine phosphate supplied as a high-purity analytical standard for research use. It is intended as a tracer or internal standard for quantitative NMR, GC-MS, and LC-MS and is provided as a white to off-white solid with recommended storage to maintain stability.

  • Molecular formula C13H14D3N6O7P.
  • Molecular weight 403.30.
  • Purity 99.68%.
  • Appearance white to off-white solid.
  • Pack size 1 MG.
  • Intended as tracer or internal standard for NMR, GC-MS, and LC-MS.
  • Storage: sealed, away from moisture; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-46341 Supplier Medchemexpress LLC Supplier No. HY10250S11MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triciribine phosphate-d3 is the deuterated isotopologue of triciribine phosphate supplied as a high-purity analytical standard for research use. It is intended as a tracer or internal standard for quantitative NMR, GC-MS, and LC-MS and is provided as a white to off-white solid with recommended storage to maintain stability.

  • Molecular formula C13H14D3N6O7P.
  • Molecular weight 403.30.
  • Purity 99.68%.
  • Appearance white to off-white solid.
  • Pack size 1 MG.
  • Intended as tracer or internal standard for NMR, GC-MS, and LC-MS.
  • Storage: sealed, away from moisture; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.