Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Triciribine phosphate sodium | 00-00-0 | 99.8% | 422.27 g/mol | C13H16N6NaO7P | 10 MG
SDP

Catalog No. 5000246561
Encompass_Preferred

Triciribine phosphate sodium is the sodium salt of triciribine (TCN-P), a small-molecule research inhibitor that affects purine metabolism and inhibits ATP synthase-related activity. Supplied as a high-purity solid, it is intended for in vitro biochemical and cell-based studies in metabolic and cancer research.

  • Sodium salt form suitable for aqueous and DMSO-based preparation.
  • High purity (~99.8%).
  • Molecular weight 422.27 g/mol.
  • Chemical formula C13H16N6NaO7P.
  • Soluble in DMSO (~62.5 mg/mL with warming or sonication).
  • Supplied as a 10 mg solid for laboratory research.
  • For research use only, not for human consumption.

Catalog No. 50-002-46561 Supplier Medchemexpress LLC Supplier No. HY10250A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triciribine phosphate sodium is the sodium salt of triciribine (TCN-P), a small-molecule research inhibitor that affects purine metabolism and inhibits ATP synthase-related activity. Supplied as a high-purity solid, it is intended for in vitro biochemical and cell-based studies in metabolic and cancer research.

  • Sodium salt form suitable for aqueous and DMSO-based preparation.
  • High purity (~99.8%).
  • Molecular weight 422.27 g/mol.
  • Chemical formula C13H16N6NaO7P.
  • Soluble in DMSO (~62.5 mg/mL with warming or sonication).
  • Supplied as a 10 mg solid for laboratory research.
  • For research use only, not for human consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.