Learn More
Cayman Chemical TrIclabendazole 100mg
SDP
Catalog No. 502984890
Encompass_Preferred
A benzimidazole anthelmintic; eliminates both immature and adult flukes in a sheep model of F. hepatica infection at 10 mg/kg; protects against glucose- or α-synuclein-induced apoptosis in S. cerevisiae at 5 µM
Catalog No. 50-298-4890 Supplier Cayman Chemical Supplier No. 18883100 MG
A benzimidazole anthelmintic; eliminates both immature and adult flukes in a sheep model of F. hepatica infection at 10 mg/kg; protects against glucose- or α-synuclein-induced apoptosis in S. cerevisiae at 5 µM