Learn More
Cayman Chemical TrIclabendazole-13C-d3 1mg
SDP
Catalog No. 502950149
Encompass_Preferred
An internal standard for the quantification of triclabendazole by GC- or LC-MS
Catalog No. 50-295-0149 Supplier Cayman Chemical Supplier No. 340111 MG
An internal standard for the quantification of triclabendazole by GC- or LC-MS