Learn More
Cayman Chemical TrIclabendazole-13C-d3 5mg
SDP
Supplier:  Cayman Chemical 340115 MG
Encompass_Preferred
An internal standard for the quantification of triclabendazole by GC- or LC-MS
Catalog No. 50-295-0150