Learn More
TARGETMOL CHEMICALS INC Triclabendazole 200MG
SDP
Catalog No. 502880552
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Triclabendazole (CGA89317) has been used in trials studying Parasitic Disease. Purity 98.35%
Catalog No. 50-288-0552 Supplier TARGETMOL CHEMICALS INC Supplier No. T0838200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Triclabendazole (CGA89317) has been used in trials studying Parasitic Disease. Purity 98.35%