Learn More
Medchemexpress LLC Triclabendazole sulfoxide | 100648-13-3 | MFCD09038493 | 98.8% | C14H9Cl3N2O2S | 10MG
SDP
Catalog No. 5000261791
Encompass_Preferred
Triclabendazole sulfoxide is the primary plasma metabolite of triclabendazole; it shows antiparasitic activity and has been reported to inhibit the ABCG2/BCRP transporter. Supplied for research use, it is typically used in pharmacology and transporter inhibition studies.
  • Main plasma metabolite of triclabendazole.
  • Exhibits antiparasitic activity.
  • Inhibits ABCG2/BCRP transporter.
  • Provided as a research-use reagent, not for human use.
  • Stable under recommended cold storage conditions for extended periods.
Catalog No. 50-002-61791 Supplier Medchemexpress LLC Supplier No. HY13645010MG
Triclabendazole sulfoxide is the primary plasma metabolite of triclabendazole; it shows antiparasitic activity and has been reported to inhibit the ABCG2/BCRP transporter. Supplied for research use, it is typically used in pharmacology and transporter inhibition studies.
  • Main plasma metabolite of triclabendazole.
  • Exhibits antiparasitic activity.
  • Inhibits ABCG2/BCRP transporter.
  • Provided as a research-use reagent, not for human use.
  • Stable under recommended cold storage conditions for extended periods.