Learn More
Medchemexpress LLC Triclabendazole sulfoxide (TCBZ-SO) | 100648-13-3 | 98.8% | 375.66 | 25 MG
SDP
Catalog No. 5000348797
Encompass_Preferred
Triclabendazole sulfoxide (TCBZ-SO) is the main plasma metabolite of Triclabendazole. It exhibits anti-parasite effects and can inhibit the membrane transporter ABCG2/BCRP.
  • Main plasma metabolite of Triclabendazole
  • Exhibits anti-parasite effects
  • Inhibits the membrane transporter ABCG2/BCRP
  • Useful in infection disease research
  • Useful in cancer disease research
  • Useful in parasitic infection research
Catalog No. 50-003-48797 Supplier Medchemexpress LLC Supplier No. 25MGHY136450
Triclabendazole sulfoxide (TCBZ-SO) is the main plasma metabolite of Triclabendazole. It exhibits anti-parasite effects and can inhibit the membrane transporter ABCG2/BCRP.
  • Main plasma metabolite of Triclabendazole
  • Exhibits anti-parasite effects
  • Inhibits the membrane transporter ABCG2/BCRP
  • Useful in infection disease research
  • Useful in cancer disease research
  • Useful in parasitic infection research