Learn More
American Radiolabeled Chemicals Inc TRICLACRYL AC 14C 50UC
SDP
Catalog No. 50353005
Encompass_Preferred
Trichloroacrylic acid-1-14C 50uCi in ethanol, 0.1 mCi/ml, 20-50 mCi/mmol, M.W. 175.4, Exclusive
Catalog No. 50-353-005 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 362050UC
Trichloroacrylic acid-1-14C 50uCi in ethanol, 0.1 mCi/ml, 20-50 mCi/mmol, M.W. 175.4, Exclusive