Learn More
Medchemexpress LLC HY-B1805 500mg Medchemexpress, Triclocarban CAS:101-20-2 Purity:>98%
SDP
Catalog No. 501933013
Encompass_Preferred
Medchemexpress, HY-B1805 500mg Triclocarban CAS:101-20-2 Triclocarban is an antimicrobial agent used in personal cleaning products. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3013 Supplier Medchemexpress LLC Supplier No. HYB1805500MG
Medchemexpress, HY-B1805 500mg Triclocarban CAS:101-20-2 Triclocarban is an antimicrobial agent used in personal cleaning products. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.