Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triclocarban, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111019593 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9593 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9593 Supplier MilliporeSigma Supelco Supplier No. 0566625MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 101-20-2
Vapor Pressure <0.1 mmHg (25°C)
Linear Formula Cl2C6H3 NHCONHC6H4 Cl
MDL Number MFCD00013254
Melting Point 254°C to 256°C
Synonym 3,4,4′-Trichlorocarbanilide; 1-(4-Chlorophenyl)-3-(3,4-dichlorophenyl)urea; TCC; Triclocarban
Type Analytical Standard
Quantity 25 mg
Formula Weight 315.58
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.