Learn More
Medchemexpress LLC Triclocarban-d4 | 1219799-29-7 | 98.0% | 1 MG
SDP
Catalog No. 5000412894
Encompass_Preferred
Triclocarban-d4 is the deuterium labeled Triclocarban. Triclocarban (3,4,4′-Trichlorocarbanilide), a broad-spectrum antibacterial compound, is widely used in applications such as the production of soaps, skin creams, toothpastes, and deodorants. It is also identified as a potential endocrine-disrupting chemical that can modulate androgen and estrogen activities, as well as other hormone-mediated biological processes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-12894 Supplier Medchemexpress LLC Supplier No. HYB1805S1MG
Triclocarban-d4 is the deuterium labeled Triclocarban. Triclocarban (3,4,4′-Trichlorocarbanilide), a broad-spectrum antibacterial compound, is widely used in applications such as the production of soaps, skin creams, toothpastes, and deodorants. It is also identified as a potential endocrine-disrupting chemical that can modulate androgen and estrogen activities, as well as other hormone-mediated biological processes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs.