Learn More
Medchemexpress LLC Triclocarban-d4 | 1219799-29-7 | 98.0% | 319.61 | C13H5D4Cl3N2O | 10 MG
SDP
Catalog No. 5000297245
Encompass_Preferred
Triclocarban-d4 is the deuterium-labeled analogue of triclocarban used as an analytical standard and internal tracer for quantitative LC-MS, GC-MS, and NMR analyses. It is supplied as a white to off-white solid intended for method development, quantitation, and metabolic or environmental studies.
  • Deuterium-labeled internal standard for quantitative LC-MS and GC-MS.
  • High purity (98.0%) suitable for analytical applications.
  • Supplied as a 10 mg solid for analytical preparation.
  • Stable isotope label enables accurate quantitation and tracer studies.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).
Catalog No. 50-002-97245 Supplier Medchemexpress LLC Supplier No. HYB1805S10MG
Triclocarban-d4 is the deuterium-labeled analogue of triclocarban used as an analytical standard and internal tracer for quantitative LC-MS, GC-MS, and NMR analyses. It is supplied as a white to off-white solid intended for method development, quantitation, and metabolic or environmental studies.
  • Deuterium-labeled internal standard for quantitative LC-MS and GC-MS.
  • High purity (98.0%) suitable for analytical applications.
  • Supplied as a 10 mg solid for analytical preparation.
  • Stable isotope label enables accurate quantitation and tracer studies.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).