Learn More
Medchemexpress LLC Triclocarban-d4 | 1219799-29-7 | 98.0% | 5 MG
SDP
Catalog No. 5000411410
Encompass_Preferred
Triclocarban-d4 is the deuterium labeled version of Triclocarban. Triclocarban (3,4,4′-Trichlorocarbanilide) is a broad-spectrum antibacterial compound commonly used in various products such as soaps, skin creams, toothpastes, and deodorants. It is also considered a potential endocrine-disrupting chemical that can modulate androgen and estrogen activities, as well as other hormone-mediated biological processes.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-11410 Supplier Medchemexpress LLC Supplier No. HYB1805S5MG
Triclocarban-d4 is the deuterium labeled version of Triclocarban. Triclocarban (3,4,4′-Trichlorocarbanilide) is a broad-spectrum antibacterial compound commonly used in various products such as soaps, skin creams, toothpastes, and deodorants. It is also considered a potential endocrine-disrupting chemical that can modulate androgen and estrogen activities, as well as other hormone-mediated biological processes.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.