Learn More
Medchemexpress LLC Triclocarban (Standard) | 101-20-2 | 99.2% | 250 MG
SDP
Catalog No. 5000413955
Encompass_Preferred
Triclocarban (Standard) is the analytical standard of Triclocarban and is intended for research and analytical applications. It is a broad-spectrum antibacterial compound widely used in products such as soaps, skin creams, toothpastes, and deodorants. This compound is also identified as a potential endocrine-disrupting chemical that can modulate androgen and estrogen activities and other hormone-mediated biological processes. The compound is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications
  • Broad-spectrum antibacterial compound
  • Potential endocrine-disrupting chemical
  • Modulates androgen and estrogen activities
  • Used in HPLC, GC, and MS experiments
Catalog No. 50-004-13955 Supplier Medchemexpress LLC Supplier No. 250MGHYB1805R
Add to Cart
Add to Cart
Triclocarban (Standard) is the analytical standard of Triclocarban and is intended for research and analytical applications. It is a broad-spectrum antibacterial compound widely used in products such as soaps, skin creams, toothpastes, and deodorants. This compound is also identified as a potential endocrine-disrupting chemical that can modulate androgen and estrogen activities and other hormone-mediated biological processes. The compound is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for research and analytical applications
  • Broad-spectrum antibacterial compound
  • Potential endocrine-disrupting chemical
  • Modulates androgen and estrogen activities
  • Used in HPLC, GC, and MS experiments