Learn More
Medchemexpress LLC Triclosan-methyl-d3 | 1020720-00-6 | 99.0% | C13H6D3Cl3O2 | 10MG
SDP
Catalog No. 5000261667
Encompass_Preferred
Triclosan-methyl-d3 is a deuterium-labeled analog of triclosan-methyl supplied as an analytical reference standard for research applications, particularly quantitative mass spectrometry and isotopic tracing. The material is provided as a solid with high purity and defined storage conditions to preserve isotopic and chemical integrity.
  • Deuterium-labeled internal standard for mass spectrometry and isotopic tracing.
  • High chemical purity (99.0%).
  • Solid form suitable for long-term storage under controlled conditions.
  • Offered in small analytical pack sizes for laboratory use.
  • Intended for research and analytical use only; not for human use.
Catalog No. 50-002-61667 Supplier Medchemexpress LLC Supplier No. HY136441S10MG
Add to Cart
Add to Cart
Triclosan-methyl-d3 is a deuterium-labeled analog of triclosan-methyl supplied as an analytical reference standard for research applications, particularly quantitative mass spectrometry and isotopic tracing. The material is provided as a solid with high purity and defined storage conditions to preserve isotopic and chemical integrity.
  • Deuterium-labeled internal standard for mass spectrometry and isotopic tracing.
  • High chemical purity (99.0%).
  • Solid form suitable for long-term storage under controlled conditions.
  • Offered in small analytical pack sizes for laboratory use.
  • Intended for research and analytical use only; not for human use.