Learn More
Cytiva TRICORN 10 COARSE FILTER KIT
Supplier:  Cytiva 11001254
Encompass
The Tricorn 10 Coarse Filter Kit is used for large volume/repeated loading of clarified sample to reduce the risk of clogging in combination with capture media.
Catalog No. 50-105-5273