Learn More
Andwin Scientific TRICORN BEAKER 100ML 100/PK
SDP
Catalog No. 50727614
Encompass_Preferred
The 100 mL Tri-Corn Beakers have feature resistance against most chemicals and temperatures up to 121C. This product also has three dripless pouring spouts and metric graduations. The beaker is measured to have a hole size of 73 mm, a diameter of 53 mm, and a height of 73 mm. It is sold in a pack of 100.
Catalog No. 50-727-614 Supplier Andwin Scientific Supplier No. 60970
Add to Cart
Add to Cart
The 100 mL Tri-Corn Beakers have feature resistance against most chemicals and temperatures up to 121C. This product also has three dripless pouring spouts and metric graduations. The beaker is measured to have a hole size of 73 mm, a diameter of 53 mm, and a height of 73 mm. It is sold in a pack of 100.