Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tricosane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015021 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5021 5 g
11-101-5020 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5021 Supplier MilliporeSigma Supelco Supplier No. 914475G
Add to Cart
Add to Cart
Petroleum
TRUSTED_SUSTAINABILITY

Specifications

CAS 638-67-5
Flash Point 113°C
Physical Form Neat
Linear Formula CH3(CH2)21 CH3
MDL Number MFCD00009350
Melting Point 46°C to 47°C; 47°C to 49°C
Boiling Point 199°C to 200°C (3 mmHg)
Type Analytical Standard
Quantity 5 g
Formula Weight 324.63
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.